RIE156328596 Rieke Metals, Inc.

1-(4-bromo-3-methylphenyl)-6-chloro-1-oxohexane

  • MDL number MFCD07700049
  • Cas Number : 898760-96-8
  • Molecular Weight : 303.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6130E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6130E
Availability details for
Quantity
  • Availability
  • Add to Cart