RIE156328597 Rieke Metals, Inc.

1-(4-bromo-3-methylphenyl)-7-chloro-1-oxoheptane

  • MDL number MFCD07700050
  • Cas Number : 898760-98-0
  • Molecular Weight : 317.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6130F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6130F
Availability details for
Quantity
  • Availability
  • Add to Cart