RIE156328598 Rieke Metals, Inc.

4-Chloro-1-(3,4-dichlorophenyl)-1-oxobutane

  • MDL number MFCD07700051
  • Cas Number : 80212-12-0
  • Molecular Weight : 251.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6131C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6131C
Availability details for
Quantity
  • Availability
  • Add to Cart