RIE156328602 Rieke Metals, Inc.

4-chloro-1-(4-fluoro-3-methylphenyl)-1-oxobutane

  • MDL number MFCD07700054
  • Cas Number : 898761-04-1
  • Molecular Weight : 214.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6133C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6133C
Availability details for
Quantity
  • Availability
  • Add to Cart