RIE156328603 Rieke Metals, Inc.

5-chloro-1-(4-fluoro-3-methylphenyl)-1-oxopentane

  • MDL number MFCD03844179
  • Cas Number : 898761-06-3
  • Molecular Weight : 228.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6133D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6133D
Availability details for
Quantity
  • Availability
  • Add to Cart