RIE156328605 Rieke Metals, Inc.

7-chloro-1-(4-fluoro-3-methylphenyl)-1-oxoheptane

  • MDL number MFCD07700056
  • Cas Number : 898761-10-9
  • Molecular Weight : 256.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6133F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6133F
Availability details for
Quantity
  • Availability
  • Add to Cart