RIE156328606 Rieke Metals, Inc.

4-chloro-1-(3-chloro-4-fluorophenyl)-1-oxobutane

  • MDL number MFCD07700057
  • Cas Number : 898761-12-1
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6134C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6134C
Availability details for
Quantity
  • Availability
  • Add to Cart