RIE156328607 Rieke Metals, Inc.

5-Chloro-1-(3-chloro-4-fluorophenyl)-1-oxopentane

  • MDL number MFCD03844180
  • Cas Number : 898761-15-4
  • Molecular Weight : 249.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6134D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6134D
Availability details for
Quantity
  • Availability
  • Add to Cart