RIE156328608 Rieke Metals, Inc.

6-chloro-1-(3-chloro-4-fluorophenyl)-1-oxohexane

  • MDL number MFCD07700058
  • Cas Number : 898761-18-7
  • Molecular Weight : 263.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6134E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6134E
Availability details for
Quantity
  • Availability
  • Add to Cart