RIE156328609 Rieke Metals, Inc.

7-chloro-1-(3-chloro-4-fluorophenyl)-1-oxoheptane

  • MDL number MFCD07700059
  • Cas Number : 898761-21-2
  • Molecular Weight : 277.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6134F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6134F
Availability details for
Quantity
  • Availability
  • Add to Cart