RIE156328617 Rieke Metals, Inc.

5-Chloro-1-(3,4-difluorophenyl)-1-oxopentane

  • MDL number MFCD03844182
  • Cas Number : 898761-51-8
  • Molecular Weight : 232.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6137D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6137D
Availability details for
Quantity
  • Availability
  • Add to Cart