RIE156328618 Rieke Metals, Inc.

6-chloro-1-(3,4-difluorophenyl)-1-oxohexane

  • MDL number MFCD07700068
  • Cas Number : 898761-54-1
  • Molecular Weight : 246.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6137E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6137E
Availability details for
Quantity
  • Availability
  • Add to Cart