RIE156328620 Rieke Metals, Inc.

4-Chloro-1-oxo-1-(4-trifluoromethoxyphenyl)butane

  • MDL number MFCD07700070
  • Cas Number : 898761-60-9
  • Molecular Weight : 266.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6138C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6138C
Availability details for
Quantity
  • Availability
  • Add to Cart