RIE156328625 Rieke Metals, Inc.

6-chloro-1-(2,4-dichlorophenyl)-1-oxohexane

  • MDL number MFCD07700074
  • Cas Number : 898786-13-5
  • Molecular Weight : 279.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6139E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6139E
Availability details for
Quantity
  • Availability
  • Add to Cart