RIE156328629 Rieke Metals, Inc.

6-chloro-1-(4-cyanophenyl)-1-oxohexane

  • MDL number MFCD07700078
  • Cas Number : 898786-19-1
  • Molecular Weight : 235.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6140E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6140E
Availability details for
Quantity
  • Availability
  • Add to Cart