RIE156328631 Rieke Metals, Inc.

5-chloro-1-oxo-1-(3-trifluoromethoxyphenyl)pentane

  • MDL number MFCD07700081
  • Cas Number : 898786-25-9
  • Molecular Weight : 280.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6141D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6141D
Availability details for
Quantity
  • Availability
  • Add to Cart