RIE156328634 Rieke Metals, Inc.

7-chloro-1-(2-methoxyphenyl)-1-oxoheptane

  • MDL number MFCD07700088
  • Cas Number : 898786-37-3
  • Molecular Weight : 254.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6142F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6142F
Availability details for
Quantity
  • Availability
  • Add to Cart