RIE156328640 Rieke Metals, Inc.

6-(2-methoxyphenyl)-6-oxohexanenitrile

  • MDL number MFCD07700098
  • Cas Number : 898786-52-2
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6145D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6145D
Availability details for
Quantity
  • Availability
  • Add to Cart