RIE156328643 Rieke Metals, Inc.

4-(3-Methoxyphenyl)-4-oxobutyronitrile

  • MDL number MFCD07700101
  • Cas Number : 38102-72-6
  • Molecular Weight : 189.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6146B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6146B
Availability details for
Quantity
  • Availability
  • Add to Cart