RIE156328647 Rieke Metals, Inc.

8-(3-methoxyphenyl)-8-oxooctanenitrile

  • MDL number MFCD07700105
  • Cas Number : 898786-64-6
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6146F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6146F
Availability details for
Quantity
  • Availability
  • Add to Cart