RIE156328649 Rieke Metals, Inc.

7-(4-methoxyphenyl)-7-oxoheptanenitrile

  • MDL number MFCD07700107
  • Cas Number : 898786-66-8
  • Molecular Weight : 231.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6147E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6147E
Availability details for
Quantity
  • Availability
  • Add to Cart