RIE156328650 Rieke Metals, Inc.

8-(4-methoxyphenyl)-8-oxooctanenitrile

  • MDL number MFCD07700108
  • Cas Number : 898786-68-0
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6147F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6147F
Availability details for
Quantity
  • Availability
  • Add to Cart