RIE156328651 Rieke Metals, Inc.

5-chloro-1-(3-cyanophenyl)-1-oxopentane

  • MDL number MFCD07700111
  • Cas Number : 898786-72-6
  • Molecular Weight : 221.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6150D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6150D
Availability details for
Quantity
  • Availability
  • Add to Cart