RIE156328652 Rieke Metals, Inc.

6-chloro-1-(3-cyanophenyl)-1-oxohexane

  • MDL number MFCD07700112
  • Cas Number : 898786-74-8
  • Molecular Weight : 235.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6150E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6150E
Availability details for
Quantity
  • Availability
  • Add to Cart