RIE156328654 Rieke Metals, Inc.

1-Chloro-5-(3,4-dimethoxyphenyl)-5-oxopentane

  • MDL number MFCD07700116
  • Cas Number : 33245-77-1
  • Molecular Weight : 256.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6153D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6153D
Availability details for
Quantity
  • Availability
  • Add to Cart