RIE156328655 Rieke Metals, Inc.

1-Chloro-6-(3,4-dimethoxyphenyl)-6-oxohexane

  • MDL number MFCD07700117
  • Cas Number : 19347-74-1
  • Molecular Weight : 270.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6153E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6153E
Availability details for
Quantity
  • Availability
  • Add to Cart