RIE156328657 Rieke Metals, Inc.

2'-Acetoxy-5-chlorovalerophenone

  • MDL number MFCD07783644
  • Cas Number : 898786-86-2
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6154D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6154D
Availability details for
Quantity
  • Availability
  • Add to Cart