RIE156328661 Rieke Metals, Inc.

3-Fluoro-4-methoxybenzoylacetonitrile

  • MDL number MFCD02260790
  • Cas Number : 898786-96-4
  • Molecular Weight : 193.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6272

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6272
Availability details for
Quantity
  • Availability
  • Add to Cart