RIE156328662 Rieke Metals, Inc.

2,4,5-Trifluorobenzoylacetonitrile

  • MDL number MFCD02260821
  • Cas Number : 142501-42-6
  • Molecular Weight : 199.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6316

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6316
Availability details for
Quantity
  • Availability
  • Add to Cart