RIE156328664 Rieke Metals, Inc.

2-Chloro-4,5-difluorobenzoylacetonitrile

  • MDL number MFCD02260826
  • Cas Number : 139218-69-2
  • Molecular Weight : 215.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6326

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6326
Availability details for
Quantity
  • Availability
  • Add to Cart