RIE156328666 Rieke Metals, Inc.

3'-Cyano-2,2,2-trifluoroacetophenone

  • MDL number MFCD01319970
  • Cas Number : 23568-85-6
  • Molecular Weight : 199.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6504-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6504-2
Availability details for
Quantity
  • Availability
  • Add to Cart