RIE156328667 Rieke Metals, Inc.

2'-Carboxy-2,2,2,-trifluoroacetophenone

  • MDL number MFCD08064170
  • Cas Number : 203124-56-5
  • Molecular Weight : 218.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6505-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6505-1
Availability details for
Quantity
  • Availability
  • Add to Cart