RIE156328668 Rieke Metals, Inc.

3'-Carboethoxy-2,2,2-trifluoroacetophenone

  • MDL number MFCD01319973
  • Cas Number : 898787-11-6
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6505-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6505-2
Availability details for
Quantity
  • Availability
  • Add to Cart