RIE156328670 Rieke Metals, Inc.

4'-n-Pentyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD01319982
  • Cas Number : 886369-31-9
  • Molecular Weight : 244.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6511D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6511D
Availability details for
Quantity
  • Availability
  • Add to Cart