RIE156328671 Rieke Metals, Inc.

2',3'-dimethyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD01319983
  • Cas Number : 886369-37-5
  • Molecular Weight : 202.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6512-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6512-1
Availability details for
Quantity
  • Availability
  • Add to Cart