RIE156328674 Rieke Metals, Inc.

2'-Methyl-2,2,2,5'-tetrafluoroacetophenone

  • MDL number MFCD01320000
  • Cas Number : 886369-93-3
  • Molecular Weight : 206.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6522

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6522
Availability details for
Quantity
  • Availability
  • Add to Cart