RIE156328675 Rieke Metals, Inc.

2'-methyl-2,2,2,4'-tetrafluoroacetophenone

  • MDL number MFCD01320001
  • Cas Number : 886370-02-1
  • Molecular Weight : 206.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6523

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6523
Availability details for
Quantity
  • Availability
  • Add to Cart