RIE156328677 Rieke Metals, Inc.

1-(4-Methylnaphthyl) trifluoromethyl ketone

  • MDL number MFCD01320010
  • Cas Number : 886370-33-8
  • Molecular Weight : 238.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6530-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6530-2
Availability details for
Quantity
  • Availability
  • Add to Cart