RIE156328680 Rieke Metals, Inc.

2'-ethyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD02260832
  • Cas Number : 204439-99-6
  • Molecular Weight : 202.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6543

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6543
Availability details for
Quantity
  • Availability
  • Add to Cart