RIE156328681 Rieke Metals, Inc.

4'-Methoxy-3'-methyl-2,2,2-trifluoroacetophenone

  • MDL number MFCD02260834
  • Cas Number : 75822-11-6
  • Molecular Weight : 218.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6546

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6546
Availability details for
Quantity
  • Availability
  • Add to Cart