RIE156328683 Rieke Metals, Inc.

2'-Chloro-4'-fluoro-2,2,2-trifluoroacetophenone

  • MDL number MFCD02260840
  • Cas Number : 886370-93-0
  • Molecular Weight : 226.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6550

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6550
Availability details for
Quantity
  • Availability
  • Add to Cart