RIE156328684 Rieke Metals, Inc.

4'-Chloro-2'-fluoro-2,2,2-trifluoroacetophenone

  • MDL number MFCD02260841
  • Cas Number : 886370-99-6
  • Molecular Weight : 226.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6551

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6551
Availability details for
Quantity
  • Availability
  • Add to Cart