RIE156328685 Rieke Metals, Inc.

2,2,2,2',4'-Pentafluoroacetophenone

  • MDL number MFCD02260842
  • Cas Number : 886371-05-7
  • Molecular Weight : 210.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6554

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6554
Availability details for
Quantity
  • Availability
  • Add to Cart