RIE156328686 Rieke Metals, Inc.

2',3'-Dichloro-2,2,2-trifluoroacetophenone

  • MDL number MFCD02260843
  • Cas Number : 886371-11-5
  • Molecular Weight : 243.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6555

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6555
Availability details for
Quantity
  • Availability
  • Add to Cart