RIE156328690 Rieke Metals, Inc.

5'-Chloro-2'-methoxy-2,2,2-trifluoroacetophenone

  • MDL number MFCD02260847
  • Cas Number : 886371-34-2
  • Molecular Weight : 238.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6559

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6559
Availability details for
Quantity
  • Availability
  • Add to Cart