RIE156328695 Rieke Metals, Inc.

3-(5-Bromo-2-methoxyphenyl)-1,1,1-trifluoro-2-propanone

  • MDL number MFCD07700129
  • Cas Number : 898787-45-6
  • Molecular Weight : 297.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6568

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6568
Availability details for
Quantity
  • Availability
  • Add to Cart