RIE156328696 Rieke Metals, Inc.

3-(3,4-Difluorophenyl)-1,1,1-trifluoro-2-propanone

  • MDL number MFCD02260856
  • Cas Number : 898787-47-8
  • Molecular Weight : 224.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6569

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6569
Availability details for
Quantity
  • Availability
  • Add to Cart