RIE156328698 Rieke Metals, Inc.

3-(2,5-Difluorophenyl)-1,1,1-trifluoro-2-propanone

  • MDL number MFCD02260858
  • Cas Number : 898787-51-4
  • Molecular Weight : 224.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6571

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6571
Availability details for
Quantity
  • Availability
  • Add to Cart