RIE156328707 Rieke Metals, Inc.

3-(4-Bromophenyl)-1,1,1-trifluoro-2-propanone

  • MDL number MFCD03844192
  • Cas Number : 161809-64-9
  • Molecular Weight : 267.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6576-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6576-3
Availability details for
Quantity
  • Availability
  • Add to Cart