RIE156328708 Rieke Metals, Inc.

3-(2-methylphenyl)-1,1,1-trifluoro-2-propanone

  • MDL number MFCD03844193
  • Cas Number : 75199-81-4
  • Molecular Weight : 202.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6577-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6577-1
Availability details for
Quantity
  • Availability
  • Add to Cart